Assay Details
Gene Symbol: march2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017006.2 | NP_001017006.1 | 3 - 4 | 493 | 82 | |
XM_018093634.1 | XP_017949123.1 | 3 - 4 | 454 | 82 | ||
GenBank mRNA | BC153690.1 | - | 3 - 4 | 601 | 82 | |
CR762288.2 | - | 3 - 4 | 493 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
