Assay Details
Gene Symbol: elac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017028.2 | NP_001017028.1 | 2 - 3 | 249 | 78 | |
GenBank mRNA | CR762044.2 | - | 2 - 3 | 249 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
