Assay Details
| Gene Symbol: mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017055.2 | NP_001017055.1 | 2 - 3 | 211 | 70 | |
| XM_012957019.2 | XP_012812473.1 | 2 - 3 | 426 | 70 | ||
| XM_012957020.2 | XP_012812474.1 | 2 - 3 | 225 | 70 | ||
| GenBank mRNA | BC121535.1 | - | 2 - 3 | 306 | 70 | |
| CR761737.2 | - | 2 - 3 | 211 | 70 | ||
More Information
Set Membership: |
|

mrps17
Genomic Map