Assay Details
| Gene Symbol: mrps30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017073.2 | NP_001017073.1 | 1 - 2 | 616 | 71 | |
| GenBank mRNA | CR761538.2 | - | 1 - 2 | 616 | 71 | |
More Information
Set Membership: |
|

mrps30
Genomic Map