Assay Details
| Gene Symbol: ppp1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017079.2 | NP_001017079.1 | 5 - 6 | 631 | 70 | |
| XM_012957029.2 | XP_012812483.1 | 5 - 6 | 700 | 70 | ||
| GenBank mRNA | BC135351.1 | - | 5 - 6 | 633 | 70 | |
| CR761503.2 | - | 5 - 6 | 631 | 70 | ||
More Information
Set Membership: |
|

ppp1r8
Genomic Map