Assay Details
Gene Symbol: med18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017086.2 | NP_001017086.1 | 2 - 3 | 247 | 92 | |
XM_018091274.1 | XP_017946763.1 | 2 - 3 | 410 | 92 | ||
GenBank mRNA | BC158934.1 | - | 2 - 3 | 368 | 92 | |
CR761425.2 | - | 2 - 3 | 247 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
