Assay Details
| Gene Symbol: bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017089.2 | NP_001017089.1 | 2 - 3 | 358 | 73 | |
| GenBank mRNA | BC158944.1 | - | 2 - 3 | 391 | 73 | |
| CR761410.2 | - | 2 - 3 | 358 | 73 | ||
More Information
Set Membership: |
|

bri3bp
Genomic Map