Assay Details
| Gene Symbol: snrpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017093.2 | NP_001017093.1 | 1 - 2 | 147 | 76 | |
| XM_012963290.1 | XP_012818744.1 | 2 - 3 | 282 | 76 | ||
| XM_012963292.1 | XP_012818746.1 | 2 - 3 | 272 | 76 | ||
| XM_012963296.1 | XP_012818750.1 | 2 - 3 | 388 | 76 | ||
| XM_018093682.1 | XP_017949171.1 | 2 - 3 | 186 | 76 | ||
| GenBank mRNA | BC077072.1 | - | 2 - 3 | 212 | 76 | |
| CR761342.2 | - | 1 - 2 | 147 | 76 | ||
More Information
Set Membership: |
|

snrpd3
Genomic Map