Assay Details
| Gene Symbol: pla2g12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017096.2 | NP_001017096.1 | 1 - 2 | 269 | 67 | |
| GenBank mRNA | BC157175.1 | - | 1 - 2 | 304 | 67 | |
| CR761315.2 | - | 1 - 2 | 269 | 67 | ||
More Information
Set Membership: |
|

pla2g12a
Genomic Map