Assay Details
Gene Symbol: tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017100.2 | NP_001017100.1 | 3 - 4 | 327 | 74 | |
XM_012957031.2 | XP_012812485.1 | 4 - 5 | 588 | 74 | ||
GenBank mRNA | CR761298.2 | - | 3 - 4 | 327 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
