Assay Details
Gene Symbol: mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017111.2 | NP_001017111.1 | 2 - 3 | 129 | 92 | |
GenBank mRNA | BC153685.1 | - | 2 - 3 | 356 | 92 | |
CR761212.2 | - | 2 - 3 | 129 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
