Assay Details
| Gene Symbol: mrpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017111.2 | NP_001017111.1 | 4 - 5 | 435 | 72 | |
| GenBank mRNA | BC153685.1 | - | 4 - 5 | 662 | 72 | |
| CR761212.2 | - | 4 - 5 | 435 | 72 | ||
More Information
Set Membership: |
|

mrpl35
Genomic Map