Assay Details
| Gene Symbol: uchl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017120.2 | NP_001017120.1 | 3 - 4 | 488 | 73 | |
| GenBank mRNA | BC135703.1 | - | 3 - 4 | 234 | 73 | |
| CR761162.2 | - | 3 - 4 | 488 | 73 | ||
More Information
Set Membership: |
|

uchl3
Genomic Map