Assay Details
Gene Symbol: spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017126.2 | NP_001017126.1 | 2 - 3 | 132 | 64 | |
XM_018092107.1 | XP_017947596.1 | 2 - 3 | 419 | 64 | ||
GenBank mRNA | BC118867.1 | - | 1 - 2 | 86 | 64 | |
BC170703.1 | - | 1 - 2 | 82 | 64 | ||
BC171111.1 | - | 1 - 2 | 82 | 64 | ||
CR761116.2 | - | 2 - 3 | 132 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
