Assay Details
Gene Symbol: comt.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017157.2 | NP_001017157.1 | 2 - 3 | 190 | 83 | |
XM_012963358.2 | XP_012818812.1 | 2 - 3 | 256 | 83 | ||
XM_012963362.2 | XP_012818816.1 | 2 - 3 | 244 | 83 | ||
GenBank mRNA | BC153689.1 | - | 1 - 2 | 164 | 83 | |
CR760877.2 | - | 2 - 3 | 190 | 83 |
Gene Symbol: LOC100135405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC121997.1 | - | 1 - 2 | 154 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
