Assay Details
Gene Symbol: gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017191.2 | NP_001017191.1 | 4 - 5 | 548 | 63 | |
GenBank mRNA | BC121552.1 | - | 4 - 5 | 467 | 63 | |
CR760599.2 | - | 4 - 5 | 548 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
