Assay Details
Gene Symbol: endog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017202.2 | NP_001017202.1 | 2 - 3 | 617 | 93 | |
GenBank mRNA | CR760515.2 | - | 2 - 3 | 617 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
