Assay Details
| Gene Symbol: glrx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017209.2 | NP_001017209.1 | 3 - 4 | 255 | 64 | |
| GenBank mRNA | BC155988.1 | - | 3 - 4 | 237 | 64 | |
| CR760465.2 | - | 3 - 4 | 255 | 64 | ||
More Information
Set Membership: |
|

glrx3
Genomic Map