Assay Details
| Gene Symbol: pitx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017227.2 | NP_001017227.1 | 1 - 2 | 742 | 103 | |
| GenBank mRNA | BC159022.1 | - | 1 - 2 | 763 | 103 | |
| CR760278.2 | - | 1 - 2 | 742 | 103 | ||
More Information
Set Membership: |
|

pitx2
Genomic Map