Assay Details
Gene Symbol: hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017234.2 | NP_001017234.1 | 7 - 8 | 604 | 55 | |
GenBank mRNA | CR760225.2 | - | 7 - 8 | 604 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
