Assay Details
Gene Symbol: lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017235.2 | NP_001017235.1 | 7 - 8 | 750 | 67 | |
XM_012964781.1 | XP_012820235.1 | 7 - 8 | 934 | 67 | ||
GenBank mRNA | CR760214.2 | - | 7 - 8 | 750 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
