Assay Details
| Gene Symbol: ndufb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017236.2 | NP_001017236.1 | 2 - 3 | 286 | 91 | |
| GenBank mRNA | BC135854.1 | - | 2 - 3 | 317 | 91 | |
| CR760211.2 | - | 2 - 3 | 286 | 91 | ||
More Information
Set Membership: |
|

ndufb7
Genomic Map