Assay Details
| Gene Symbol: ezh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017293.2 | NP_001017293.1 | 7 - 8 | 838 | 68 | |
| XM_012964796.2 | XP_012820250.1 | 8 - 9 | 1102 | 68 | ||
| XM_012964797.2 | XP_012820251.1 | 8 - 9 | 1083 | 68 | ||
| XM_012964798.2 | XP_012820252.1 | 7 - 8 | 947 | 68 | ||
| XM_018094676.1 | XP_017950165.1 | 8 - 9 | 1104 | 68 | ||
| XM_018094677.1 | XP_017950166.1 | 8 - 9 | 1085 | 68 | ||
| GenBank mRNA | CR855647.2 | - | 7 - 8 | 838 | 68 | |
More Information
Set Membership: |
|

ezh2
Genomic Map