Assay Details
Gene Symbol: prrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017345.2 | NP_001017345.1 | 3 - 4 | 238 | 83 | |
XM_012957065.2 | XP_012812519.1 | 4 - 5 | 417 | 83 | ||
XM_018091288.1 | XP_017946777.1 | 4 - 5 | 524 | 83 | ||
GenBank mRNA | CR855488.2 | - | 3 - 4 | 238 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
