Assay Details
Gene Symbol: cdc42se2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030323.2 | NP_001025494.1 | 4 - 5 | 759 | 64 | |
GenBank mRNA | BC123961.1 | - | 3 - 4 | 443 | 64 | |
CR762307.2 | - | 4 - 5 | 759 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
