Assay Details
Gene Symbol: cdc42se2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030333.2 | NP_001025504.1 | 3 - 4 | 396 | 80 | |
GenBank mRNA | CR761398.2 | - | 3 - 4 | 396 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
