Assay Details
Gene Symbol: tfap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037258.1 | NP_001032335.1 | 5 - 6 | 965 | 67 | |
XM_012964477.2 | XP_012819931.1 | 5 - 6 | 1855 | 67 | ||
XM_012964478.2 | XP_012819932.1 | 5 - 6 | 1794 | 67 | ||
XM_012964480.2 | XP_012819934.1 | 5 - 6 | 1075 | 67 | ||
XM_012964481.2 | XP_012819935.1 | 5 - 6 | 1548 | 67 | ||
XM_012964482.2 | XP_012819936.1 | 5 - 6 | 1062 | 67 | ||
XM_012964483.2 | XP_012819937.1 | 5 - 6 | 1014 | 67 | ||
XM_012964484.2 | XP_012819938.1 | 5 - 6 | 1487 | 67 | ||
XM_018094599.1 | XP_017950088.1 | 5 - 6 | 952 | 67 | ||
XM_018094600.1 | XP_017950089.1 | 6 - 7 | 1163 | 67 | ||
GenBank mRNA | AF176818.1 | - | 5 - 6 | 879 | 67 | |
BC135698.1 | - | 5 - 6 | 1348 | 67 | ||
CR848274.2 | - | 5 - 6 | 1028 | 67 | ||
CR848276.2 | - | 5 - 6 | 965 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
