Assay Details
| Gene Symbol: ctsc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037259.1 | NP_001032336.1 | 1 - 2 | 178 | 63 | |
| XM_012956689.2 | XP_012812143.1 | 1 - 2 | 256 | 63 | ||
| GenBank mRNA | BC067941.1 | - | 1 - 2 | 204 | 63 | |
| BC098085.1 | - | 1 - 2 | 173 | 63 | ||
| CR761110.2 | - | 1 - 2 | 178 | 63 | ||
More Information
Set Membership: |
|

ctsc
Genomic Map