Assay Details
| Gene Symbol: clns1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037262.1 | NP_001032339.1 | 1 - 2 | 156 | 60 | |
| GenBank mRNA | BC087765.1 | - | 1 - 2 | 132 | 60 | |
| BC157253.1 | - | 1 - 2 | 150 | 60 | ||
| CR761131.2 | - | 1 - 2 | 156 | 60 | ||
More Information
Set Membership: |
|

clns1a
Genomic Map