Assay Details
Gene Symbol: tubb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037268.1 | NP_001032345.1 | 335 | 84 | ||
GenBank mRNA | BC090613.1 | - | 335 | 84 | ||
CR942335.2 | - | 3 - 4 | 351 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
