Assay Details
| Gene Symbol: akr7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039732.1 | NP_001034821.1 | 593 | 76 | ||
| GenBank mRNA | BC076912.1 | - | 584 | 76 | ||
| BC171130.1 | - | 3 - 4 | 585 | 76 | ||
| CR759999.2 | - | 593 | 76 | |||
More Information
Set Membership: |
|

akr7a2
Genomic Map