Assay Details
Gene Symbol: tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039737.1 | NP_001034826.1 | 7 - 8 | 828 | 98 | |
GenBank mRNA | BC088774.1 | - | 7 - 8 | 673 | 98 | |
BC118714.1 | - | 7 - 8 | 716 | 98 | ||
CR761337.2 | - | 7 - 8 | 828 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
