Assay Details
| Gene Symbol: tnnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203626.2 | NP_988957.1 | 2 - 3 | 126 | 79 | |
| XM_012971592.2 | XP_012827046.1 | 2 - 3 | 154 | 79 | ||
| GenBank mRNA | BC061286.1 | - | 2 - 3 | 133 | 79 | |
| CR760837.2 | - | 2 - 3 | 126 | 79 | ||
More Information
Set Membership: |
|

tnnc2
Genomic Map