Assay Details
| Gene Symbol: hmgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203732.2 | NP_989063.2 | 2 - 3 | 204 | 65 | |
| XM_012956648.2 | XP_012812102.1 | 2 - 3 | 294 | 65 | ||
| XM_018091141.1 | XP_017946630.1 | 2 - 3 | 158 | 65 | ||
| XM_018091142.1 | XP_017946631.1 | 2 - 3 | 479 | 65 | ||
| GenBank mRNA | BC061309.1 | - | 2 - 3 | 217 | 65 | |
| CR760370.2 | - | 2 - 3 | 204 | 65 | ||
| CR760897.2 | - | 2 - 3 | 130 | 65 | ||
| CR761959.2 | - | 2 - 3 | 203 | 65 | ||
| CR848284.2 | - | 2 - 3 | 209 | 65 | ||
More Information
Set Membership: |
|

hmgn1
Genomic Map