Assay Details
Gene Symbol: grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203937.3 | NP_989268.3 | 3 - 4 | 550 | 73 | |
GenBank mRNA | BC064215.1 | - | 3 - 4 | 542 | 73 | |
CR760996.2 | - | 3 - 4 | 533 | 73 | ||
CX515001.2 | - | 3 - 4 | 551 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
