Assay Details
Gene Symbol: rgs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203964.3 | NP_989295.2 | 3 - 4 | 388 | 69 | |
GenBank mRNA | BC064234.1 | - | 3 - 4 | 295 | 69 | |
CN080459.1 | - | 3 - 4 | 325 | 69 | ||
CR926434.1 | - | 3 - 4 | 369 | 69 | ||
CX876971.1 | - | 3 - 4 | 388 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
