Assay Details
Gene Symbol: apex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203976.2 | NP_989307.1 | 2 - 3 | 317 | 84 | |
XM_012958392.2 | XP_012813846.1 | 3 - 4 | 425 | 84 | ||
XM_012958402.2 | XP_012813856.1 | 3 - 4 | 403 | 84 | ||
XM_012958408.2 | XP_012813862.1 | 3 - 4 | 403 | 84 | ||
XM_012958414.2 | XP_012813868.1 | 3 - 4 | 403 | 84 | ||
GenBank mRNA | BC064266.1 | - | 2 - 3 | 323 | 84 | |
BC080950.1 | - | 2 - 3 | 321 | 84 | ||
CR760104.2 | - | 2 - 3 | 317 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
