Assay Details
Gene Symbol: rab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004787.2 | NP_001004787.1 | 2 - 3 | 211 | 71 | |
GenBank mRNA | BC074522.1 | - | 2 - 3 | 364 | 71 | |
BC127356.1 | - | 2 - 3 | 211 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
