Assay Details
| Gene Symbol: rpl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004911108.3 | XP_004911165.1 | 3 - 4 | 261 | 81 | |
| XM_012955556.2 | XP_012811010.1 | 3 - 4 | 199 | 81 | ||
| GenBank mRNA | BC077039.1 | - | 1 | 207 | 81 | |
| CR942419.2 | - | 183 | 81 | |||
| CT025306.2 | - | 1 | 183 | 81 | ||
| DN100063.1 | - | 2 - 3 | 238 | 81 | ||
| DT440085.1 | - | 2 - 3 | 236 | 81 | ||
More Information
Set Membership: |
|

rpl34
Genomic Map