Assay Details
| Gene Symbol: nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007489.2 | NP_001007490.1 | 3 - 4 | 485 | 69 | |
| GenBank mRNA | BC079927.1 | - | 3 - 4 | 510 | 69 | |
| CR942453.2 | - | 3 - 4 | 485 | 69 | ||
More Information
Set Membership: |
|

nfe2l2
Genomic Map