Assay Details
| Gene Symbol: gna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008053.2 | NP_001008054.1 | 1 - 2 | 535 | 64 | |
| GenBank mRNA | BC080940.1 | - | 1 - 2 | 361 | 64 | |
| CR942615.2 | - | 1 - 2 | 535 | 64 | ||
More Information
Set Membership: |
|

gna14
Genomic Map