Assay Details
| Gene Symbol: gna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008053.2 | NP_001008054.1 | 3 - 4 | 870 | 78 | |
| GenBank mRNA | BC080940.1 | - | 3 - 4 | 696 | 78 | |
| CR942615.2 | - | 3 - 4 | 870 | 78 | ||
More Information
Set Membership: |
|

gna14
Genomic Map