Assay Details
Gene Symbol: alg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016192.3 | NP_001016192.2 | 3 - 4 | 433 | 120 | |
GenBank mRNA | BC123914.1 | - | 3 - 4 | 433 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
