Assay Details
| Gene Symbol: dglucy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016461.3 | NP_001016461.2 | 11 - 12 | 1773 | 102 | |
| GenBank mRNA | BC123974.1 | - | 11 - 12 | 1773 | 102 | |
| CR760948.2 | - | 12 - 13 | 1819 | 102 | ||
More Information
Set Membership: |
|

dglucy
Genomic Map