Assay Details
Gene Symbol: trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016642.3 | NP_001016642.1 | 3 - 4 | 294 | 85 | |
XM_012956993.2 | XP_012812447.1 | 3 - 4 | 459 | 85 | ||
GenBank mRNA | BC121995.1 | - | 3 - 4 | 294 | 85 | |
CR760400.2 | - | 2 - 3 | 284 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
