Assay Details
Gene Symbol: retreg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016672.3 | NP_001016672.2 | 7 - 8 | 981 | 73 | |
XM_012954518.2 | XP_012809972.1 | 8 - 9 | 948 | 73 | ||
XM_012954519.2 | XP_012809973.1 | 8 - 9 | 945 | 73 | ||
XM_018097745.1 | XP_017953234.1 | 7 - 8 | 921 | 73 | ||
GenBank mRNA | BC121834.1 | - | 7 - 8 | 981 | 73 | |
CR760241.2 | - | 7 - 8 | 831 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
