Assay Details
Gene Symbol: atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017310.3 | NP_001017310.1 | 7 - 8 | 903 | 84 | |
GenBank mRNA | BC125665.1 | - | 7 - 8 | 903 | 84 | |
CR855501.2 | - | 7 - 8 | 915 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
