Assay Details
| Gene Symbol: echdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044397.1 | NP_001037862.1 | 2 - 3 | 261 | 72 | |
| GenBank mRNA | BC088780.1 | - | 1 - 2 | 179 | 72 | |
| BC118874.1 | - | 1 - 2 | 197 | 72 | ||
| BC170562.1 | - | 1 - 2 | 239 | 72 | ||
| BC170919.1 | - | 1 - 2 | 239 | 72 | ||
| CR942395.2 | - | 2 - 3 | 261 | 72 | ||
More Information
Set Membership: |
|

echdc1
Genomic Map