Assay Details
| Gene Symbol: atp5po | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044412.1 | NP_001037877.1 | 1 - 2 | 143 | 79 | |
| GenBank mRNA | BC121272.1 | - | 1 - 2 | 143 | 79 | |
| CR760022.2 | - | 1 - 2 | 40 | 79 | ||
More Information
Set Membership: |
|

atp5po
Genomic Map