Assay Details
Gene Symbol: nthl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044419.1 | NP_001037884.1 | 2 - 3 | 340 | 70 | |
GenBank mRNA | BC159395.1 | - | 2 - 3 | 388 | 70 | |
CR760124.2 | - | 2 - 3 | 340 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
